Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LCMT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583898
Description
Immunogen sequence: TLVFPSSEAF PRVNPASPSG VFPASVVSSE GQVPNLKRYG HASVFLSPDV ILSAGGFGEQ EGRHCRVSQF HLLSRDCDSE WKGSQIGSCG TGVQWDGR.
TYW4 is an enzyme that participates in the wybutosine-tRNA (Phe) biosynthesis pathway. Wybutosine (yW) is a hypermodified guanosine at the 3-prime position adjacent to the anticodon of phenylalanine tRNA that stabilizes codon-anticodon interactions during decoding on the ribosome. TYW4 is involved in a multistep enzymatic reaction that stabilizes codon-anticodon base-pairing during the ribosomal decoding process, thereby ensuring correct translation.
Specifications
LCMT2 | |
Polyclonal | |
Unconjugated | |
LCMT2 | |
D330024M17; KIAA0547; LCMT2; Leucine carboxyl methyltransferase 2; p21WAF1/CIP1 promoter-interacting protein; PPM2; tRNA wybutosine-synthesizing protein 4; tRNA wybutosine-synthesizing protein 4 homolog; tRNA yW-synthesizing protein 4; tRNA yW-synthesizing protein 4 homolog; tRNA(Phe) (7-(3-amino-3-(methoxycarbonyl)propyl)wyosine(37)-N)-methoxycarbonyltransferase; tRNA(Phe) (7-(3-amino-3-carboxypropyl)wyosine(37)-O)-methyltransferase; tRNA-Wybutosine-synthesizing protein 4; tRNA-yW synthesizing protein 4; TYW4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9836 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60294 | |
LCMT2 | |
Recombinant protein corresponding to Human LCMT2. Recombinant protein control fragment (Product #RP-104849). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction