Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LCMT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521155
Description
A suggested positive control is rat brain tissue lysate. PA5-21155 can be used with blocking peptide PEP-1269.
TYW4 is an enzyme that participates in the wybutosine-tRNA (Phe) biosynthesis pathway. Wybutosine (yW) is a hypermodified guanosine at the 3-prime position adjacent to the anticodon of phenylalanine tRNA that stabilizes codon-anticodon interactions during decoding on the ribosome. TYW4 is involved in a multistep enzymatic reaction that stabilizes codon-anticodon base-pairing during the ribosomal decoding process, thereby ensuring correct translation.
Specifications
LCMT2 | |
Polyclonal | |
Unconjugated | |
LCMT2 | |
D330024M17; KIAA0547; LCMT2; Leucine carboxyl methyltransferase 2; p21WAF1/CIP1 promoter-interacting protein; PPM2; tRNA wybutosine-synthesizing protein 4; tRNA wybutosine-synthesizing protein 4 homolog; tRNA yW-synthesizing protein 4; tRNA yW-synthesizing protein 4 homolog; tRNA(Phe) (7-(3-amino-3-(methoxycarbonyl)propyl)wyosine(37)-N)-methoxycarbonyltransferase; tRNA(Phe) (7-(3-amino-3-carboxypropyl)wyosine(37)-O)-methyltransferase; tRNA-Wybutosine-synthesizing protein 4; tRNA-yW synthesizing protein 4; TYW4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
296098, 329504, 9836 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O60294, Q5XIA3, Q8BYR1 | |
LCMT2 | |
A 16 amino acid peptide near the amino terminus of human TYW4. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction