Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCN1 Monoclonal Antibody (10B10)
GREENER_CHOICE

Catalog No. PIMA515501
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515501 Supplier Invitrogen™ Supplier No. MA515501
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15501 targets LCN1 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15501 immunogen is purified recombinant fragment of LCN1 expressed in E. Coli.. MA5-15501 detects LCN1 which has a predicted molecular weight of approximately 19.2kDa.

The protein encoded by this gene belongs to the lipocalin family. Lipocalins are a group of extracellular proteins that are able to bind lipophiles by enclosure within their structures to minimize solvent contact. This protein may bind hydrophobic ligands and inhibit cysteine proteinases. It may also play a role in taste reception.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCN1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10B10
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene LCN1
Gene Accession No. P31025
Gene Alias LCN1; lipocalin 1; lipocalin 1 (tear prealbumin); lipocalin-1; MGC71975; PMFA; protein migrating faster than albumin; Tear lipocalin; Tear prealbumin; Tlc; TP; VEG protein; VEGP; von Ebner gland protein
Gene Symbols LCN1
Host Species Mouse
Immunogen Purified recombinant fragment of LCN1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3933
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.