Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCN1 Monoclonal Antibody (7H4H6)
GREENER_CHOICE

Catalog No. PIMA530633
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530633 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530633 Supplier Invitrogen™ Supplier No. MA530633
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human LCN1/Lipocalin 1.

The protein encoded by this gene belongs to the lipocalin family. Lipocalins are a group of extracellular proteins that are able to bind lipophiles by enclosure within their structures to minimize solvent contact. This protein may bind hydrophobic ligands and inhibit cysteine proteinases. It may also play a role in taste reception.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCN1
Applications ELISA
Classification Monoclonal
Clone 7H4H6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene LCN1
Gene Accession No. P31025
Gene Alias LCN1; lipocalin 1; lipocalin 1 (tear prealbumin); lipocalin-1; MGC71975; PMFA; protein migrating faster than albumin; Tear lipocalin; Tear prealbumin; Tlc; TP; VEG protein; VEGP; von Ebner gland protein
Gene Symbols LCN1
Host Species Mouse
Immunogen Recombinant Human LCN1/Lipocalin 1 Protein (Met1-Asp176).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3933
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.