Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCN10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563207 Supplier Invitrogen™ Supplier No. PA563207
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GHLWASLSVK GVKAFHVLST DYSYGLVYLR LGRATQNYKN LLLFHRQNVS SFQSLKEFMD ACDILGLSKA AVILPKDASR THTI Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 65%.

May play a role in male fertility. May act as a retinoid carrier protein within the epididymis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCN10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LCN10
Gene Accession No. Q6JVE6
Gene Alias 9230112J07Rik; Epididymal-specific lipocalin-10; Lcn10; lipocalin 10
Gene Symbols LCN10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LCN10. Recombinant protein control fragment (Product #RP-99972).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 414332
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.