Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LCN15 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306604MFV500
Description
LCN15 Polyclonal antibody specifically detects LCN15 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| LCN15 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| lipocalin 15, lipocalin-15, MSFL2541, PRO6093, UNQ2541 | |
| Produced in rabbits immunized with E. coli-derived Human LCN15 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 389812 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction