Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCN15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563882 Supplier Invitrogen™ Supplier No. PA563882
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FAVLYIYKEL EGALSTMVQL YSRTQDVSPQ ALKAFQDFYP TLGLPEDMMV MLPQSDACNP ESKE Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 36%.

The LCN15 (Lipocalin 15) gene encodes a protein that belongs to the lipocalin family, a group of small extracellular proteins characterized by their ability to bind and transport small hydrophobic molecules. LCN15 is a human-specific protein found abundantly in the olfactory mucus. It has been identified as a biomarker for Bowman gland cells in the olfactory neuroepithelium, with its expression correlating with non-degenerated areas of this tissue. The lipocalin family proteins, including LCN15, exhibit significant sequence heterogeneity but possess highly conserved tertiary structures with an eight-stranded antiparallel β-barrel. This structure forms a binding pocket that allows them to transport a wide variety of molecules, including retinol, steroids, and various lipids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCN15
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LCN15
Gene Accession No. Q6UWW0
Gene Alias LCN15; lipocalin 15; lipocalin-15; MSFL2541; PRO6093; UNQ2541; UNQ2541/PRO6093
Gene Symbols LCN15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LCN15. Recombinant protein control fragment (Product #RP-101526).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389812
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.