Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCN2 Goat anti-Rat, Polyclonal , Abnova™

Catalog No. 89934463 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-463 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-463 Supplier Abnova Corporation Supplier No. PAB29709

Goat polyclonal antibody raised against rat Lcn2.

Specifications

Antigen LCN2
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Gene Symbols Lcn2
Host Species Goat
Immunogen Recombinant protein corresponding to rat Lcn2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170496
Target Species Rat
Content And Storage Store at 4°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.