Learn More
LCN2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | LCN2 |
---|---|
Concentration | 300 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8546
![]() ![]() |
Proteintech Group Inc
269911AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The LCN2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human LCN2. This antibody recognizes human antigen. The LCN2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
LCN2 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human | |
IgG |
300 μg/mL | |
Polyclonal | |
Rabbit | |
3934 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.