Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCN6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102384
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102384 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102384 Supplier Invitrogen™ Supplier No. PA5102384
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LCN6.

LCN6 is a member of the lipocalin family of hydrophobic ligand binding proteins and exhibit conserved beta strand basket like structures. LCN6 may play a role in male fertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCN6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LCN6
Gene Accession No. P62502
Gene Alias 9230101D24Rik; epididymal-specific lipocalin ELP16; epididymal-specific lipocalin LCN6; epididymal-specific lipocalin-6; hLcn5; LCN5; Lcn6; lipocalin 5; lipocalin 6; lipocalin-5; MCG18768; UNQ643; UNQ643/PRO1273
Gene Symbols LCN6
Host Species Rabbit
Immunogen A synthesized peptide derived from human LCN6(Accession P62502), corresponding to amino acid residues A48-E98.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158062, 414138, 620709
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.