Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCN8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984184
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4184 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4184 Supplier Bioss Supplier No. BS18196R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in male fertility. May act as a retinoid carrier protein within the epididymis.

Specifications

Antigen LCN8
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LCN8
Gene Accession No. Q6JVE9
Gene Alias EP17; Epididymal-specific lipocalin-8; LCN5; LCN8; lipocalin 5; lipocalin 8
Gene Symbols LCN8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LCN8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138307
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.