Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCN9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584473 Supplier Invitrogen™ Supplier No. PA584473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQEFDPHTVM QRNYNVARVS GVWYSIFMAS DDLNRIKENG DLRVFVRNIE HLKNGSLIFD FEYMVQ Highest antigen sequence indentity to the following orthologs: Rat - 48%, Mouse - 56%.

Members of the lipocalin family, such as LCN9, have a common structure consisting of an 8-stranded antiparallel beta-barrel that forms a cup-shaped ligand-binding pocket or calyx. Lipocalins generally bind small hydrophobic ligands and transport them to specific cells (Suzuki et al., 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCN9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LCN9
Gene Accession No. Q8WX39
Gene Alias 9230102I19Rik; Epididymal-specific lipocalin-9; epididymis luminal protein 129; HEL129; LCN9; lipocalin 9; MUP-like lipocalin
Gene Symbols LCN9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LCN9. Recombinant protein control fragment (Product #RP-108449).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 392399
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.