Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCOR Monoclonal Antibody (731229)
GREENER_CHOICE

Catalog No. PIMA524345
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524345 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524345 Supplier Invitrogen™ Supplier No. MA524345
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

LCOR is a transcriptional corepressor widely expressed in fetal and adult tissues that is recruited to agonist-bound nuclear receptors through a single LxxLL motif, also referred to as a nuclear receptor (NR) box.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCOR
Applications Immunocytochemistry
Classification Monoclonal
Clone 731229
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene LCOR
Gene Accession No. Q96JN0
Gene Alias 3110023F06Rik; A630025C20Rik; C10orf12; KIAA1795; Lcor; ligand dependent nuclear receptor corepressor; ligand-dependent corepressor; Mblk1-related protein 2; Mblk1-related protein-2; mKIAA1795; MLR2; RGD1311704; transcription factor MLR2
Gene Symbols LCOR
Host Species Mouse
Immunogen E. coli-derived recombinant human LCoR Asn110-Gln338.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84458
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.