Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCOR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584354 Supplier Invitrogen™ Supplier No. PA584354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSEEGNTCII PQRNLFKALS EEAWNSGFMG NSSRTADKEN TLQCPKTPLR QDLEANEQDA RPKQENHLHS LGRNKVGYHL HPSDKGQFD Highest antigen sequence indentity to the following orthologs: Rat - 70%, Mouse - 72%.

LCOR is a transcriptional corepressor widely expressed in fetal and adult tissues that is recruited to agonist-bound nuclear receptors through a single LxxLL motif, also referred to as a nuclear receptor (NR) box.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCOR
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LCOR
Gene Accession No. Q96JN0
Gene Alias 3110023F06Rik; A630025C20Rik; C10orf12; KIAA1795; Lcor; ligand dependent nuclear receptor corepressor; ligand-dependent corepressor; Mblk1-related protein 2; Mblk1-related protein-2; mKIAA1795; MLR2; RGD1311704; transcription factor MLR2
Gene Symbols LCOR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LCOR. Recombinant protein control fragment (Product #RP-108426).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84458
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.