Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCORL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568896 Supplier Invitrogen™ Supplier No. PA568896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

May act as transcription activator that binds DNA elements with the sequence 5'-CCCTATCGATCGATCTCTACCT-3'. May play a role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCORL
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LCORL
Gene Accession No. Q8N3X6
Gene Alias fi43c07; im:7138788; Lcorl; LCoR-like protein; LCORLp1; LCORLp2; ligand dependent nuclear receptor corepressor like; ligand dependent nuclear receptor corepressor-like; ligand-dependent nuclear receptor corepressor-like protein; MBLK1-related protein; mblk1-related protein 1; Mblk1-related protein-1; MLR1; RGD1561241; si:dkeyp-26a9.6; Transcription factor MLR1; wu:fi43c07
Gene Symbols LCORL
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse LCORL.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254251
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.