Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LCT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144271
Description
LCT Polyclonal Antibody for IHC (P), ELISA

Specifications
LCT | |
Polyclonal | |
Unconjugated | |
LCT | |
LAC; lactase; Lactase-glycosylceramidase; lactase-phlorizin hydrolase; lactase-phlorizin hydrolase-1; LCT; LPH; LPH1; Phlorizin hydrolase | |
Rabbit | |
Protein G | |
RUO | |
3938 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
4.99 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P09848 | |
LCT | |
Recombinant Human Lactase-phlorizin hydrolase protein (983-1252AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction