Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCTL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567189 Supplier Invitrogen™ Supplier No. PA567189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SIWDVFTHSGK GKVLGNETAD VACDGYYKVQ EDIILLRELH VNHYRFSLSW Highest antigen sequence identity to the following orthologs - mouse 75%, rat 71%.

This gene encodes a member of family 1 glycosidases. Glycosidases are enzymes that hydrolyze glycosidic bonds and are classified into families based on primary amino acid sequence. Most members of family 1 have two conserved glutamic acid residues, which are required for enzymatic activity. The mouse ortholog of this protein has been characterized and has a domain structure of an N-terminal signal peptide, glycosidase domain, transmembrane domain, and a short cytoplasmic tail. It lacks one of the conserved glutamic acid residues important for catalysis, and its function remains to be determined. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCTL
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LCTL
Gene Accession No. Q6UWM7
Gene Alias KL lactase phlorizin hydrolase; KLG; klotho gamma; klotho/lactase-phlorizin hydrolase-related protein; KLPH; lactase like; lactase-like; lactase-like protein; LCTL; UNQ3022/PRO9820
Gene Symbols LCTL
Host Species Rabbit
Immunogen Recombinant Human LCTL. Recombinant protein control fragment (Product #RP-107829).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197021
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.