Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDAH Antibody (OTI2G9), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272122B
Description
LDAH Monoclonal specifically detects LDAH in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
LDAH | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
C2ORF43 | |
Full length recombinant protein of human C2orf43 (NP_068744) produced in HEK293T cells. | |
0.1 mL | |
60526.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
OTI2G9 | |
Western Blot, Flow Cytometry | |
chromosome 2 open reading frame 43, FLJ21820, hypothetical protein LOC60526, lipid droplet associated hydrolase | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction