Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDB1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131362
Description
LDB1 Polyclonal specifically detects LDB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
LDB1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 | |
LDB1 | |
Recombinant protein encompassing a sequence within the center region of human LDB1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
8861 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1.01mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
carboxy terminal LIM domain protein 2, Carboxyl-terminal LIM domain-binding protein 2, CLIM-2, CLIM2hLdb1, LDB-1, LIM domain binding 1, LIM domain-binding factor-1, LIM domain-binding protein 1, NLILIM domain-binding factor CLIM2, Nuclear LIM interactor | |
Rabbit | |
47 kDa | |
0.1 mL | |
Primary | |
Tested in human and Mouse. Expected to cross react based on sequence identity: Rhesus Monkey (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction