Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDB1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572653
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572653 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572653 Supplier Invitrogen™ Supplier No. PA572653
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LDB1A Polyclonal Antibody for Western Blot

Binds to the LIM domain of a wide variety of LIM domain-containing transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDB1A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ldb1a
Gene Accession No. O73715
Gene Alias ldb1a; LDB-1-A; ldb4; LDB-4; ldb4 protein; ldb4b; LIM domain binding 1a; LIM domain-binding protein 1-A; LIM domain-binding protein 4; LIM-domain binding factor 1a; LIM-domain binding factor 4; LIM-domain binding factor 4b; zLdb4
Gene Symbols ldb1a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 336-368 amino acids from the C-terminal region of DANRE ldb1a.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30579
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.