Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDB2 Polyclonal Antibody
LDB2 Polyclonal Antibody
Supplier: Thermo Scientific PA531346
Description
LDB2 Polyclonal Antibody for Western Blot

Specifications
LDB2 | |
Polyclonal | |
Unconjugated | |
LDB2 | |
AI035351; AW146358; Carboxyl-terminal LIM domain-binding protein 1; cLdb2; Clim1; CLIM-1; CLP-36; LDB1; Ldb2; LDB-2; Ldb3; LIM binding domain 2; LIM domain binding 2; LIM domain binding protein CLIM-1; LIM domain-binding factor CLIM1; LIM domain-binding factor-2; LIM domain-binding protein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16826, 9079 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.73 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O43679, O55203 | |
LDB2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 235 of Human LDB2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction