Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100276
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100276 Supplier Invitrogen™ Supplier No. PA5100276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LDB3.

This gene encodes a PDZ domain-containing protein. PDZ motifs are modular protein-protein interaction domains consisting of 80-120 amino acid residues. PDZ domain-containing proteins interact with each other in cytoskeletal assembly or with other proteins involved in targeting and clustering of membrane proteins. The protein encoded by this gene interacts with alpha-actinin-2 through its N-terminal PDZ domain and with protein kinase C via its C-terminal LIM domains. The LIM domain is a cysteine-rich motif defined by 50-60 amino acids containing two zinc-binding modules. This protein also interacts with all three members of the myozenin family. Mutations in this gene have been associated with myofibrillar myopathy and dilated cardiomyopathy. Alternatively spliced transcript variants encoding different isoforms have been identified; all isoforms have N-terminal PDZ domains while only longer isoforms (1, 2 and 5) have C-terminal LIM domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDB3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LDB3
Gene Accession No. O75112, Q9JKS4
Gene Alias AW742271; cardiomyopathy, dilated 1C (autosomal dominant); CMD1C; CMH24; CMPD3; cypher; FLJ35865; HGNC:15710; KIAA01613; KIAA0613; LDB3; LDB3Z1; LDB3Z4; LIM domain binding 3; LIM domain-binding protein 3; LVNC3; MFM4; ORACLE; oracle protein; PDLIM6; PDZ and LIM domain 6; PDZ-LIM domain protein; protein cypher; Protein oracle; RGD1564875; ZASP; Z-band alternatively spliced PDZ motif protein; Z-band alternatively spliced PDZ-motif protein
Gene Symbols LDB3
Host Species Rabbit
Immunogen A synthesized peptide derived from human LDB3(Accession O75112), corresponding to amino acid residues T522-P572.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11155, 24131, 498587
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.