Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDHA Mouse anti-Human, Biotin, Clone: OTI2B11, liquid, TrueMAB™
SDP

Catalog No. 501677581
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7581 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7581 Supplier OriGene Supplier No. TA700008
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lactate dehydrogenase (LDH) is the enzyme in the glycolytic pathway that converts pyruvate to lactate with concomitant interconversion of NADH and NAD+. In mammals, the enzyme is encoded by three genes: LDHA (M or muscle form), LDHB (H or heart form), and LDHC (X or testis form). The three human LDHs have 84-89% sequence similarities and 69-75% amino acid identities. There are five different LDH isoenzymes based on the proportion of M and H chains existing in the LDH tetrameric structure. The LDHA is hypoxia inducible and its expression is directly controlled by the transcriptional activity of the hypoxia inducible factor 1a (HIF1a). LDHB is also known to be upregulated in many cancers.

Specifications

Antigen LDHA
Applications ELISA
Classification Monoclonal
Clone OTI2B11
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 10mg/ml BSA, 50% glycerol and 0.05% sodium azide
Gene LDHA
Gene Accession No. P00338
Gene Alias GSD11, HEL-S-133P, LDHM, PIG19
Gene Symbols LDHA
Host Species Mouse
Immunogen Protein expressed in 293T cell transfected with human LDHA expression vector
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3939
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.