Learn More
LDHA Mouse anti-Human, Clone: OTI2C11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500518

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Lactate dehydrogenase (LDH) is the enzyme in the glycolytic pathway that converts pyruvate to lactate with concomitant interconversion of NADH and NAD+. In mammals, the enzyme is encoded by three genes: LDHA (M or muscle form), LDHB (H or heart form), and LDHC (X or testis form). The three human LDHs have 84-89% sequence similarities and 69-75% amino acid identities. There are five different LDH isoenzymes based on the proportion of M and H chains existing in the LDH tetrameric structure. The LDHA is hypoxia inducible and its expression is directly controlled by the transcriptional activity of the hypoxia inducible factor 1a (HIF1a). LDHB is also known to be upregulated in many cancers.Specifications
LDHA | |
Monoclonal | |
Unconjugated | |
LDHA | |
GSD11, HEL-S-133P, LDHM, PIG19 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3939 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
OTI2C11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P00338 | |
LDHA | |
Full length human recombit protein of human LDHA produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.