Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDHA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579593
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579593 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579593 Supplier Invitrogen™ Supplier No. PA579593
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, human MCF-7 whole cell, human A375 whole cell, human HepG2 whole cell, rat liver tissue, rat RH35 whole cell, mouse liver tissue. IHC: mouse intestine tissue, rat intestine tissue, human mammary cancer tissue, mouse skeletal tissue, rat muscle skeletal tissue, human stomach cancer tissue, human thyroid cancer tissue, human pancreas cancer tissue. ICC/IF: U20S cell. Flow: A549 cell.

Lactate dehydrogenase (LDH) is the enzyme in the glycolytic pathway that converts pyruvate to lactate with concomitant interconversion of NADH and NAD+. In mammals, the enzyme is encoded by three genes: LDHA (M or muscle form), LDHB (H or heart form), and LDHC (X or testis form). The three human LDHs have 84-89% sequence similarities and 69-75% amino acid identities. There are five different LDH isoenzymes based on the proportion of M and H chains existing in the LDH tetrameric structure. The LDHA is hypoxia inducible and its expression is directly controlled by the transcriptional activity of the hypoxia inducible factor 1a (HIF1a). LDHB is also known to be upregulated in many cancers. Mutations in the LDHA gene can lead to Glycogen storage disease 11 (GSD11).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDHA
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene LDHA
Gene Accession No. P00338, P04642, P06151, P19629
Gene Alias Cell proliferation-inducing gene 19 protein; epididymis secretory sperm binding protein Li 133P; GSD11; HEL-S-133P; l7R2; Lactate Dehydrogenase; lactate dehydrogenase 1, A chain; lactate dehydrogenase 3, C chain, sperm specific; lactate dehydrogenase A; lactate dehydrogenase A4; lactate dehydrogenase A-like; lactate dehydrogenase C; lactate dehydrogenase C variant 3; lactate dehydrogenase M; lactate dehydrogenase-A; lactate dehydrogenase-M; lactate dehydrogenase-M (EC 1.1.1.27); Lactic Dehydrogenase; LDH; LDH muscle subunit; LDH testis subunit; Ldh1; Ldh-1; Ldh3; Ldha; LDH-A; ldha protein; ldha1; LDH-A4 muscle-specific isozyme; glycolytic enzyme; Ldhc; LDH-C; Ldhm; LDH-M; LDH-X; L-lactate dehydrogenase; L-lactate dehydrogenase A chain; L-lactate dehydrogenase C chain; M-LDH; OTTHUMP00000165206; PIG19; proliferation-inducing gene 19; renal carcinoma antigen NY-REN-59
Gene Symbols LDHA
Host Species Rabbit
Immunogen E. coli-derived human LDHA recombinant protein (Position: A2-R106).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16828, 24533, 29634, 3939
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.