Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ LDHA, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN896 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN896 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN896 Supplier MilliporeSigma™ Supplier No. ABN896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

L-lactate dehydrogenase A chain (EC 1.1.1.27; UniProt P00338; also known as Cell proliferation-inducing gene 19 protein, LDH muscle subunit, LDH-A, LDH-M, Renal carcinoma antigen NY-REN-59) is encoded by the LDHA (also known as GSD11, PIG19) gene (Gene ID 3939) in human. Originally described by Otto Warburg and now commonly known as the Warburg effect refers to the phenomenon that many tumor cells use aerobic glycolysis instead of mitochondrial respiration for energy production to support cell growth and proliferation. Enzymes involved in the complex network of metabolic pathways are therefore being studied as potential targets for cancer treatment. Lactate dehydrogenase (LDH) is a tetrameric enzyme composed of two different subunits, LDHA and LDHB, which can assemble into five different combinations (LDH1 is comprised of four LDHB subunits; LDH2 contains three LDHB and one LDHA subunit; LDH3 contains two LDHB and two LDHA subunits; LDH4 contains one LDHB and three LDHA subunits; LDH5 contains four LDHA subunits). LDHB is ubiquitously expressed and the predominant isoform in heart muscle, while LDHA is the predominant isoform in skeletal muscle and other highly glycolytic tissues. LDH catalyzes the reversible conversion of pyruvate into lactate. An upregulation of LDH therefore reduces the oxygen dependency and ensures an efficient anaerobic/glycolytic metabolism in tumor cells. Upregulated LDHA has been observed in hepatocellular carcinoma, breast, pancreatic, and prostate cancers. Inhibition of LDHA reduced cell malignant transformation and tumor formation. Serum LDHB level is also reported to elevate in lung cancer patients and progressively increase with the clinical stage of non-small cell lung cancer (NSCLC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDHA
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene LDHA, GSD11, PIG19
Gene Accession No. P00338
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a C-terminal region sequence of human LDHA.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_005557
Test Specificity This rabbit polyclonal targeted epitope region is present in human LDHA spliced isoforms 1, 3, and 4, but absent from isoforms 2 and 5, reported by UniProt (P00338).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.