Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDHAL6A Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5100667

Catalog No. PIPA5100667


Only null left
Add to Cart

Description

Description

Antibody detects endogenous levels of total LDHAL6A.

Displays an lactate dehydrogenase activity. Significantly increases the transcriptional activity of JUN, when overexpressed.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LDHAL6A
Polyclonal
Unconjugated
LDHAL6A
lactate dehydrogenase A like 6A; lactate dehydrogenase A-like 6A; LDH6A; LDHAL6A; LDHL2; L-lactate dehydrogenase A-like 6A
Rabbit
Affinity chromatography
RUO
160287
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6ZMR3
LDHAL6A
A synthesized peptide derived from human LDHAL6A(Accession Q6ZMR3), corresponding to amino acid residues L214-L264.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.