Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LDHAL6A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524372
Description
LDHAL6A Polyclonal Antibody for Western Blot
Displays an lactate dehydrogenase activity. Significantly increases the transcriptional activity of JUN, when overexpressed.
Specifications
| LDHAL6A | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6ZMR3 | |
| LDHAL6A | |
| KLH conjugated synthetic peptide between 154-183 amino acids from the central region of human LDHAL6A. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| LDHAL6A | |
| lactate dehydrogenase A like 6A; lactate dehydrogenase A-like 6A; LDH6A; LDHAL6A; LDHL2; L-lactate dehydrogenase A-like 6A | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 160287 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction