Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDHAL6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569998 Supplier Invitrogen™ Supplier No. PA569998
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Human: 100%; Pig: 79%; Rabbit: 79%; Rat: 77%.

LDHAL6B belongs to the LDH/MDH superfamily. LDH family. The specific function of the protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDHAL6B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LDHAL6B
Gene Accession No. Q9BYZ2
Gene Alias 4933402O15Rik; AI326310; lactate dehydrogenase A like 6B; lactate dehydrogenase A-like 6; lactate dehydrogenase A-like 6B; LDH6B; Ldhal; LDHAL6; Ldhal6b; LDHL; L-lactate dehydrogenase A-like 6B; testicular tissue protein Li 105
Gene Symbols LDHAL6B
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human LDHAL6B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92483
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.