Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDHB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596736 Supplier Invitrogen™ Supplier No. PA596736
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATLKEKLIA PVAEEEATVP NNKITVVGVG QVGMACAISI LGKSLADELA LVDVLEDKLK GEMMDLQHGS LFLQTPKIVA DKDYSVTANS KIVVVTAGVR QQEGESRLNL VQRNVNVFKF IIPQIVKYSP DCIIIVVSNP VDILTYVTWK LSGLPKHRVI GSGCNLDSAR FRYLMAEKLG IHPSSCHGWI LGEHGDSSVA VWSGVNVAGV SLQELNPEMG TDNDSENWKE VHKMVVESAY EVIKLKGYTN WAIGLSVADL IESMLKNLSR IHPVSTMVKG MYGIENEVFL SLPCILNARG LTSVINQKLK DDEVAQLKKS ADTLWDIQKD LKDL; Positive Samples: HeLa, A-431, Jurkat, A-549, Mouse brain, Mouse kidney, Mouse heart, Mouse lung, Rat brain; Cellular Location: Cytoplasm.

This gene encodes an enzyme which catalyzes the reversible conversion of lactate and pyruvate, and NAD and NADH, in the glycolytic pathway. Mutations in this gene are associated with lactate dehydrogenase B deficiency. Pseudogenes have been identified on the X chromosome and on chromosome. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDHB
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene LDHB
Gene Accession No. P07195, P16125, P42123
Gene Alias AI790582; epididymis secretory protein Li 281; H(B)-type lactate dehydrogenase; HEL-S-281; H-Ldh; lactate dehydrogenase B; lactate dehydrogenase H chain; lactate dehydrogenase-B; lactate dehydrogenase-H; lactate dehydrogenase-H (, EC 1.1.1.27); Lactate dehydrogenease B; LDH heart subunit; Ldh2; Ldh-2; LDHB; LDH-B; LDHBD; LDHH; LDH-H; L-lactate dehydrogenase B chain; Renal carcinoma antigen NY-REN-46; similar to lactate dehydrogenase B; testicular secretory protein Li 25; TRG-5
Gene Symbols LDHB
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16832, 24534, 3945
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.