Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LDHD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559716
Description
Immunogen sequence: PWRGYCSQKA KGELCRDFVE ALKAVVGGSH VSTAAVVREQ HGRDESVHRC EPPDAVVWPQ NVEQVSRLAA LCYRQGVPII PFGTGTGLEG GVCAVQG Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.
LDHD belongs to the D-isomer specific 2-hydroxyacid dehydrogenase family. The similar protein in yeast has both D-lactate and D-glycerate dehydrogenase activities. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
LDHD | |
Polyclonal | |
Unconjugated | |
LDHD | |
4733401P21Rik; Ac2-202; D8Bwg1320e; D-lactate dehydrogenase; DLD; lactate dehydrogenase D; LDHD; Probable D-lactate dehydrogenase, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
197257 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86WU2 | |
LDHD | |
Recombinant protein corresponding to Human LDHD. Recombinant protein control fragment (Product #RP-98713). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction