Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDHD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598588
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598588 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598588 Supplier Invitrogen™ Supplier No. PA598588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LDHD Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

LDHD belongs to the D-isomer specific 2-hydroxyacid dehydrogenase family. The similar protein in yeast has both D-lactate and D-glycerate dehydrogenase activities. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDHD
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LDHD
Gene Accession No. Q86WU2
Gene Alias 4733401P21Rik; Ac2-202; D8Bwg1320e; D-lactate dehydrogenase; DLD; lactate dehydrogenase D; LDHD; Probable D-lactate dehydrogenase, mitochondrial
Gene Symbols LDHD
Host Species Rabbit
Immunogen Recombinant Human Probable D-lactate dehydrogenase, mitochondrial protein (310-507AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197257
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.