Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ LDL Receptor, Mouse, Unlabeled, Clone: 2H7.1,

Catalog No. MABS1208MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1208MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1208MI Supplier MilliporeSigma™ Supplier No. MABS1208
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Low-density lipoprotein receptor (UniProt P01130; also known as LDL receptor) is encoded by the LDLR (also known as FH, FHC) gene (Gene ID 3949) in human. Low-density lipoprotein (LDL) receptor recognizes the apoprotein B100 embedded in the outer phospholipid layer of cholesterol-rich LDL particles and mediates their remval by endocytosis via clathrin-coated pits. LDL receptor also recognizes the apoE protein found in chylomicron remnants and VLDL remnants (IDL). LDLR gene mutations are known causes of familial hypercholesterolaemia (FH). LDL receptor is initially produced with a signal peptide sequence (a.a. 1-21), the removal of which yields the mature single transmembrane (a.a. 789-810) protein wiht a C-terminal cytoplasmic tail (a.a. 811-860) and a large N-terminal extracellular region (a.a. 22-788) that contains seven class A (a.a. 25-313) and six class B (a.a. 397-658) LDLR repeats, as well as three EGF-like domains (a.a. 314-353, 354-393, 663-712). LDL-A repeats mediate LDL binding, repeats 2-7 are required for ApoB binding, while only repeat 5 is required for ApoE binding. The class B (LDL-B) repeats region adopts a beta-propeller conformation and is believed to be responsible for the pH-dependent release of LDL in the endosome. The cytoplasmic tail contains a signal sequence important for localizing the receptors to clathrin-coated pits and for triggering receptor-mediated endocytosis upon LDL binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDL Receptor
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2H7.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene LDLR, FH, FHC
Gene Accession No. P01130
Host Species Mouse
Immunogen GST-tagged recombinant human LDL receptor extracellular fragment corresponding to class A domains 1-3.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000518
Test Specificity Expected to react with spliced isoforms 1 and 5 reported by UniProt P01130). Reactivity toward spliced isoforms 2, 3, 4, and 6 has not been determined.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.