Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDLR Antibody (11A2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | LDLR |
---|---|
Clone | 11A2 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:10-1:20, ELISA 1:100-2000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP222035
![]() |
Novus Biologicals
NBP222035 |
0.1 mg | N/A | N/A | N/A | ||||
Description
LDLR Monoclonal specifically detects LDLR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
LDLR | |
LYOPH | |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
Unconjugated | |
Mouse | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
LDLR | |
IgG1 | |
Protein G purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
11A2 | |
Western Blot 1:500-1:1000, Flow Cytometry 1:10-1:20, ELISA 1:100-2000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P01130 | |
3949 | |
Human recombinant LDLR | |
Primary | |
Reconstitute antibody with 500ul of sterile PBS to a concentration of 0.2mg/ml. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title