Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDLR Antibody (C7), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP178159APC
Description
LDLR Monoclonal specifically detects LDLR in Human, Rat, Porcine, Bovine samples. It is validated for Flow Cytometry.Specifications
LDLR | |
Monoclonal | |
APC | |
P01130 | |
LDLR | |
Partially purified bovine LDL receptor [Swiss-Prot# P01131]. | |
0.1 mL | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
3949 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
C7 | |
Flow Cytometry | |
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Rat, Pig, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction