Learn More
LDLR Antibody (C7), PE, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP178159PE
Catalog No. NBP178159PE
Only null left
Add to Cart
Description
Description
LDLR Monoclonal specifically detects LDLR in Human, Rat, Porcine, Bovine samples. It is validated for Flow Cytometry.
Specifications
Specifications
LDLR
Monoclonal
PE
P01130
LDLR
Partially purified bovine LDL receptor [Swiss-Prot# P01131].
0.1 mL
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
3949
Store at 4C in the dark.
IgG2b
Flow Cytometry
C7
Flow Cytometry
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Mouse
Protein G purified
RUO
Primary
Human, Rat, Pig, Bovine
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only