Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDLR Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP106709PE
Description
LDLR Polyclonal antibody specifically detects LDLR in Human, Mouse samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDownSpecifications
LDLR | |
Polyclonal | |
Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated | |
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
Synthetic peptide made to an internal portion of the human LDL Receptor protein (within residues 500-550). [Swiss-Prot# P01130] | |
0.1 mL | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
3949 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction