Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDLR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522976 Supplier Invitrogen™ Supplier No. PA522976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with monkey based on 100% sequence homology. This antibody has 92% sequence homology with mouse.

The low density lipoprotein receptor (LDL-R) gene family consists of cell surface proteins involved in receptor-mediated endocytosis of specific ligands. Low density lipoprotein (LDL) is normally bound at the cell membrane and taken into the cell ending up in lysosomes where the protein is degraded and the cholesterol is made available for repression of microsomal enzyme 3-hydroxy-3-methylglutaryl coenzyme A (HMG CoA) reductase, the rate-limiting step in cholesterol synthesis. At the same time, a reciprocal stimulation of cholesterol ester synthesis takes place. Mutations in the LDL-R gene cause the autosomal dominant disorder, familial hypercholesterolemia. Along with SCARB1, CLDN1, and the tetraspanin superfamily member CD81, LDL-R has been reported to be an entry factor for the Hepatitis C virus. At least three isoforms of LDL-R are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDLR
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene LDLR
Gene Accession No. P01130, P35951
Gene Alias FH; FHC; Hlb301; I79_005860; LDL receptor; LDLA; LDLCQ2; LDLR; ldl-r; LDLRA; low density lipoprotein receptor; low density lipoprotein receptor protein; low-density lipoprotein receptor; low-density lipoprotein receptor class A domain-containing protein 3
Gene Symbols LDLR
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human LDL Receptor protein (within residues 500-550).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16835, 3949
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.