Learn More
LDLR Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304417A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine and Rabbit
The low density lipoprotein (LDL) receptor system coordinates the metabolism of cholesterol, an essential component of the plasma membrane of all mammalian cells. Study of this system has led to an enhanced understanding of the cellular basis of cholesterol homeostasis. It has also brought into focus an important mechanism of metabolic regulation--the process of receptor-mediated endocytosis (1). Data suggest that the juxtamembranous region of the cytoplasmic domain participates in protein:protein interactions that allow the low density lipoprotein receptor to cluster in coated pits (2). It has been shown that the family of LDL receptors may serve as viral receptors. Endocytosis of the Flaviviridae viruses, hepatitis C virus, GB virus C/hepatitis G virus, and bovine viral diarrheal virus (BVDV) was shown to be mediated by LDL receptors on cultured cells (3).Specifications
LDLR | |
Polyclonal | |
Unconjugated | |
LDLR | |
familial hypercholesterolemia, FH, FHC, LDL receptor, LDLCQ2, low-density lipoprotein receptor class A domain-containing protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3949 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P01130 | |
LDLR | |
Between 810 and 860 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.