Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDLR Rabbit anti-Mouse, Clone: 101, Novus Biologicals™

Rabbit Monoclonal Antibody
$366.00 - $584.50
Specifications
Antigen | LDLR |
---|---|
Clone | 101 |
Dilution | Immunocytochemistry/Immunofluorescence 1:20-1:100 |
Applications | Immunofluorescence |
Classification | Monoclonal |
Description
LDLR Monoclonal specifically detects LDLR in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
LDLR | |
Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
Recombinant Mouse LDLR Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
101 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
0.2 um filtered solution in PBS | |
3949 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title