Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDLR Rabbit anti-Mouse, Clone: 32, Novus Biologicals™

Rabbit Monoclonal Antibody
$414.50 - $584.50
Specifications
Antigen | LDLR |
---|---|
Clone | 32 |
Dilution | Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:1000 |
Applications | Flow Cytometry, ELISA, Immunofluorescence |
Classification | Monoclonal |
Description
LDLR Monoclonal specifically detects LDLR in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
LDLR | |
Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
Recombinant Mouse LDLR protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
32 | |
Flow Cytometry, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
0.2 um filtered solution in PBS | |
3949 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title