Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDLR Recombinant Rabbit Monoclonal Antibody (10K4L8)
GREENER_CHOICE

Catalog No. FEMA551226
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551226 Supplier Invitrogen™ Supplier No. MA551226
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: TVEIVTMSHQ ALGDVAGRGN EKKPSSVRAL SIVLPIVLLV FLCLGVFLLW KNWRLKNINS INFDNPVYQK TTEDEVHICH NQDGYSYPSR QMVSLEDDVA.

The low density lipoprotein receptor (LDL-R) gene family consists of cell surface proteins involved in receptor-mediated endocytosis of specific ligands. Low density lipoprotein (LDL) is normally bound at the cell membrane and taken into the cell ending up in lysosomes where the protein is degraded and the cholesterol is made available for repression of microsomal enzyme 3-hydroxy-3-methylglutaryl coenzyme A (HMG CoA) reductase, the rate-limiting step in cholesterol synthesis. At the same time, a reciprocal stimulation of cholesterol ester synthesis takes place. Mutations in the LDL-R gene cause the autosomal dominant disorder, familial hypercholesterolemia. Along with SCARB1, CLDN1, and the tetraspanin superfamily member CD81, LDL-R has been reported to be an entry factor for the Hepatitis C virus. At least three isoforms of LDL-R are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDLR
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 10K4L8
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene LDLR
Gene Accession No. P01130, P35951, P35952
Gene Alias FH; FHC; Hlb301; I79_005860; LDL receptor; LDLA; LDLCQ2; LDLR; ldl-r; LDLRA; low density lipoprotein receptor; low density lipoprotein receptor protein; low-density lipoprotein receptor; low-density lipoprotein receptor class A domain-containing protein 3
Gene Symbols LDLR
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16835, 300438, 3949
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.