Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDLRAD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101654 Supplier Invitrogen™ Supplier No. PA5101654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LDLRAD2.

The exact functions of this protein remain unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDLRAD2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LDLRAD2
Gene Accession No. Q5SZI1
Gene Alias LDLRAD2; low density lipoprotein receptor A domain containing 2; low density lipoprotein receptor class A domain containing 2; low-density lipoprotein receptor class A domain-containing protein 2; lrad2
Gene Symbols LDLRAD2
Host Species Rabbit
Immunogen A synthesized peptide derived from human LDLRAD2(Accession Q5SZI1), corresponding to amino acid residues P129-C179.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401944
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.