Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDLRAP1 Monoclonal Antibody (OTI2B2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503840S


Description
LDLRAP1 Monoclonal antibody specifically detects LDLRAP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
LDLRAP1 | |
Monoclonal | |
0.54 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q5SW96 | |
Mouse | |
Affinity chromatography | |
RUO | |
26119 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI2B2 | |
Unconjugated | |
LDLRAP1 | |
AA691260; Arh; Arh1; ARH2; Autosomal recessive hypercholesterolemia protein; autosomal recessive hypercholesterolemia protein homolog; DKFZp586D0624; FHCB1; FHCB2; LDL receptor adaptor protein; LDLRAP1; Low density lipoprotein receptor adapter protein 1; low density lipoprotein receptor adaptor protein 1; MGC34705; RGD1563417; RP11-70P17.2 | |
Full length human recombinant protein of human LDLRAP1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction