Learn More
LDLRAP1 Mouse anti-Human, Clone: OTI4B10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503846

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a cytosolic protein which contains a phosphotyrosine binding (PTD) domain. The PTD domain has been found to interact with the cytoplasmic tail of the LDL receptor. Mutations in this gene lead to LDL receptor malfunction and cause the disorder autosomal recessive hypercholesterolaemia.Specifications
LDLRAP1 | |
Monoclonal | |
Unconjugated | |
LDLRAP1 | |
ARH, ARH1, ARH2, FHCB1, FHCB2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
26119 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI4B10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q5SW96 | |
LDLRAP1 | |
Full length human recombit protein of human LDLRAP1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.