Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LDN 193188
GREENER_CHOICESDP

Catalog No. 563150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
563150 50 mg
563110 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 563150 Supplier Tocris Bioscience™ Supplier No. 5631/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of phosphatidylcholine transfer protein (PC-TP)

LDN 193188 is an inhibitor of phosphatidylcholine transfer protein (PC-TP). Regulates hepatic glucose metabolism. Also promotes insulin-independent phosphorylation of key insulin signaling molecules.

Specifications

Chemical Name or Material 2,4-Dichloro-N-[[[4-[[(4,6-dimethyl-2-pyrimidinyl)amino]sulfonyl]phenyl]amino]carbonyl]benzamide
CAS 1267610-30-9
Quantity 50 mg
Target Lipid Metabolism Inhibitors
Molecular Formula C20H17Cl2N5O4S
Formula Weight 494.35
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.