Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LDN 193189 dihydrochloride
GSA_VAGREENER_CHOICE

Catalog No. 605310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-531-0 10 mg
60-535-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-531-0 Supplier Tocris Bioscience™ Supplier No. 6053/10
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Potent and selective ALK2 and ALK3 inhibitor; inhibits BMP4 signaling; promotes neural induction of hPSCs

LDN 193189 dihydrochloride is a potent and selective ALK2 and ALK3 inhibitor (IC50 values are 5 and 30 nM, respectively); inhibits BMP4-mediated Smad1/5/8 activation. Exhibits >200-fold selectivity for BMP signaling over TGF-β signaling. Also exhibits selectivity over AMPK, PDGFR and MAPK signaling. Promotes neural induction of hPSCs in combination with SB 431542 (Cat.No. 1614). Also induces differentiation of hPSCs into nociceptive sensory neurons in combination with SB 431542 (Cat.No. 1614), SU 5402 (Cat.No.3300), CHIR 99021 (Cat.No. 4423) and DAPT (Cat.No. 2634).

LDN 193189 synthesized to Ancillary Material Grade also available.

For more information about how LDN 193189 dihydrochloride may be used, see our protocols: Generation of β cells from hPSCs, Generating Midbrain Dopaminergic Neurons from hPSCs, Accelerated Induction of Cortical Neurons from hiPSCs .

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Chemical Name or Material 4-[6-[4-(1-Piperazinyl)phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]quinoline dihydrochloride
CAS 1435934-00-1
Synonym 4-[6-[4-(1-Piperazinyl)phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]quinoline dihydrochloride
Quantity 10 mg
Solubility Soluble to 50mM in water and to 20mM in DMSO
Target BMP and Other Activin Receptor Inhibitors
Molecular Formula C25H22N60.2 HCl
Formula Weight 479.4
Physical Form Solid
Color Orange
Product Type Bioactive Small Molecule
Purity 98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.