Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDOC1 Antibody (OTI1A2), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271763UV
Description
LDOC1 Monoclonal specifically detects LDOC1 in Human samples. It is validated for Western Blot.Specifications
LDOC1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LDOC1 | |
Full length recombinant protein of human LDOC1 (NP_036449) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI1A2 | |
Western Blot | |
BCUR1, breast cancer, up-regulated 1, Leucine zipper protein down-regulated in cancer cells, leucine zipper, down-regulated in cancer 1, Mar7, Mart7, protein LDOC1 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
23641.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction