Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDOC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566429 Supplier Invitrogen™ Supplier No. PA566429
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISLLTGEAEEW VVPYIEMDSP ILGDYRAFLD EMKQCFGWDD DEDDD Highest antigen sequence identity to the following orthologs - mouse 72%, rat 81%.

LDOC1 contains a leucine zipper-like motif and a proline-rich region that shares marked similarity with an SH3-binding domain. The protein localizes to the nucleus and is down-regulated in some cancer cell lines. It is thought to regulate the transcriptional response mediated by the nuclear factor kappa B (NF-kappaB). The gene has been proposed as a tumor suppressor gene whose protein product may have an important role in the development and/or progression of some cancers.The protein encoded by this gene contains a leucine zipper-like motif and a proline-rich region that shares marked similarity with an SH3-binding domain. The protein localizes to the nucleus and is down-regulated in some cancer cell lines. It is thought to regulate the transcriptional response mediated by the nuclear factor kappa B (NF-kappaB). The gene has been proposed as a tumor suppressor gene whose protein product may have an important role in the development and/or progression of some cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDOC1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ldoc1
Gene Accession No. O95751
Gene Alias 7-Mar; BCUR1; breast cancer, up-regulated 1; Gm366; Ldoc1; LDOC1 regulator of NFKB signaling; LDOC1, regulator of NFKB signaling; Leucine zipper; leucine zipper downregulated in cancer; leucine zipper down-regulated in cancer 1; leucine zipper protein down-regulated in cancer cells; leucine zipper, down-regulated in cancer 1; Mar7; Mart7; Protein LDOC1; RGD1565644
Gene Symbols Ldoc1
Host Species Rabbit
Immunogen Recombinant Human LDOC1. Recombinant protein control fragment (Product #RP-107103).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23641
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.