Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDOC1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599234 Supplier Invitrogen™ Supplier No. PA599234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LDOC1L.

LDOC1L gene ontology (GO) annotation includes protein binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDOC1L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RTL6
Gene Accession No. Q505G4, Q6ICC9
Gene Alias 6-Mar; BC058638; dJ1033E15.2; gag-like protein; LDOC1L; leucine zipper down-regulated in cancer 1 like; leucine zipper protein down-regulated in cancer cells-like; leucine zipper, down-regulated in cancer 1 like; leucine zipper, down-regulated in cancer 1-like; mammalian retrotransposon-derived protein 6; MAR6; MART6; protein LDOC1L; Protein Sushi-15E3; Retrotransposon Gag-like protein 6; Rtl6; Sushi-15E3
Gene Symbols RTL6
Host Species Rabbit
Immunogen A synthesized peptide derived from human RTL6(Accession Q6ICC9), corresponding to amino acid residues E149-V199.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223732, 84247
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.